Assay Details
Target Gene Details
Entrez Gene ID: | 8975 |
Gene Name: | ubiquitin specific peptidase 13 (isopeptidase T-3) |
Gene Aliases: |
ISOT3, IsoT-3 |
Location: |
Chr.3:179653145-179789401 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP13 | NM_003940.2 | NP_003931.2 | ||
| XM_011513269.1 | XP_011511571.1 | |||
| XM_017007425.1 | XP_016862914.1 | |||
| XM_017007426.1 | XP_016862915.1 | |||
| AK290338.1 | ||||
| AK302404.1 | ||||
| BC016146.1 | AAH16146.1 | |||
| HM138083.1 | ||||
| U75362.1 | AAC63405.1 |
More Information
Set Membership: |
|

Genomic Map