Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Gene Details
Entrez Gene ID: | 100874244 |
Gene Name: | NAALADL2 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:175235092-175271096 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAALADL2-AS2 | NR_046713.1 | |||
| BQ049755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829800 | Chr.3:175252630 - 175424883 on Build GRCh38 | Gain+Loss |
|
| nsv461021 | Chr.3:175143245 - 175354813 on Build GRCh38 | Loss |
|
| esv2762367 | Chr.3:175268370 - 175286432 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map