Assay Details
Target Gene Details
Entrez Gene ID: | 55076 |
Gene Name: | transmembrane protein 45A |
Gene Aliases: |
DERP7, DNAPTP4 |
Location: |
Chr.3:100492619-100577453 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM45A | NM_018004.2 | NP_060474.1 | ||
| XM_017006715.1 | XP_016862204.1 | |||
| AK000996.1 | BAA91458.1 | |||
| AK129789.1 | ||||
| AY450392.1 | AAR21085.1 |

Genomic Map