Assay Details
Target Gene Details
Entrez Gene ID: | 6304 |
Gene Name: | SATB homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.3:18345388-18445588 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATB1 | NM_001131010.3 | 11 | 5970 | NP_001124482.1 |
| NM_001195470.2 | 12 | 7568 | NP_001182399.1 | |
| NM_001322871.1 | 12 | 6114 | NP_001309800.1 | |
| NM_001322872.1 | 12 | 6018 | NP_001309801.1 | |
| NM_001322873.1 | 11 | 6018 | NP_001309802.1 | |
| NM_001322874.1 | 11 | 5832 | NP_001309803.1 | |
| NM_001322875.1 | 11 | 5830 | NP_001309804.1 | |
| NM_001322876.1 | 10 | 5783 | NP_001309805.1 | |
| NM_002971.5 | 11 | 7472 | NP_002962.1 | |
| AK124253.1 | 1 | 1800 | ||
| DB347409.1 | 1 | 338 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589852 | Chr.3:18318496 - 18345841 on Build GRCh38 | Loss |
|
| nsv589853 | Chr.3:18344240 - 18386958 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map