Assay Details
Target Gene Details
Entrez Gene ID: | 25937 |
Gene Name: | WW domain containing transcription regulator 1 |
Gene Aliases: |
TAZ |
Location: |
Chr.3:149517235-149724783 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWTR1 | NM_001168278.1 | NP_001161750.1 | ||
| XM_017006121.1 | XP_016861610.1 | |||
| XM_017006122.1 | XP_016861611.1 | |||
| AK022036.1 | BAB13957.1 | |||
| DA338432.1 |
Target Gene Details
Entrez Gene ID: | 100128025 |
Gene Name: | WWTR1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:149658210-149661364 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWTR1-AS1 | NR_040250.1 | |||
| AK130903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829754 | Chr.3:149654237 - 149798267 on Build GRCh38 | Gain |
|
| esv34142 | Chr.3:149632394 - 150056429 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map