Assay Details
Target Gene Details
Entrez Gene ID: | 100507524 |
Gene Name: | ARHGEF26 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:154024401-154121210 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF26-AS1 | NR_037901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598247 | Chr.3:154034789 - 154036121 on Build GRCh38 | Loss |
|

Genomic Map