Assay Details
Target Gene Details
Entrez Gene ID: | 100506724 |
Gene Name: | long intergenic non-protein coding RNA 901 |
Gene Aliases: |
LSAMP-AS4, TCONS_00005428 |
Location: |
Chr.3:116921431-116932238 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00901 | NR_121607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160972 | Chr.3:116855358 - 116930532 on Build GRCh38 | Duplication |
|
| esv3597499 | Chr.3:116789478 - 116999700 on Build GRCh38 | Loss |
|
| nsv1011135 | Chr.3:116380194 - 119036472 on Build GRCh38 | Gain |
|
| nsv1004313 | Chr.3:116918089 - 117061175 on Build GRCh38 | Loss |
|

Genomic Map