Assay Details
Target Gene Details
Entrez Gene ID: | 56987 |
Gene Name: | BBX, HMG-box containing |
Gene Aliases: |
ARTC1, HBP2, HSPC339, MDS001 |
Location: |
Chr.3:107522936-107811329 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591266 | Chr.3:107385775 - 107957486 on Build GRCh38 | Gain |
|
| nsv829654 | Chr.3:107435249 - 107587384 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map