Assay Details
Target Gene Details
Entrez Gene ID: | 152078 |
Gene Name: | PQ loop repeat containing 2 like |
Gene Aliases: |
C3orf55 |
Location: |
Chr.3:157543344-157601233 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PQLC2L | NM_001130001.2 | 4 | 1262 | NP_001123473.1 |
| NM_001130002.2 | 5 | 1351 | NP_001123474.1 | |
| BC029129.1 | 5 | 1298 | AAH29129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4081 | Chr.3:157600677 - 157629866 on Build GRCh38 | Deletion |
|
| nsv829767 | Chr.3:157588622 - 157772655 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map