Assay Details
Target Gene Details
Entrez Gene ID: | 5290 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha |
Gene Aliases: |
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha |
Location: |
Chr.3:179148114-179240093 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CA | NM_006218.3 | NP_006209.2 | ||
| XM_006713658.3 | XP_006713721.1 | |||
| XM_011512894.2 | XP_011511196.1 | |||
| AK292940.1 | ||||
| BX484734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592649 | Chr.3:178944026 - 179168821 on Build GRCh38 | Gain |
|
| nsv1003540 | Chr.3:179158015 - 179168505 on Build GRCh38 | Loss |
|
| nsv1010571 | Chr.3:179159177 - 179170365 on Build GRCh38 | Gain |
|
| nsv1161006 | Chr.3:179154246 - 179174436 on Build GRCh38 | Duplication |
|
| nsv822351 | Chr.3:179160729 - 179166016 on Build GRCh38 | Loss |
|
| esv2726256 | Chr.3:179160833 - 179166083 on Build GRCh38 | Deletion |
|
| esv2659099 | Chr.3:179160825 - 179166095 on Build GRCh38 | Deletion |
|
| esv3598835 | Chr.3:179160798 - 179166176 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map