Assay Details
Target Gene Details
Entrez Gene ID: | 57552 |
Gene Name: | neutral cholesterol ester hydrolase 1 |
Gene Aliases: |
AADACL1, NCEH |
Location: |
Chr.3:172630645-172711218 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCEH1 | NM_001146276.1 | NP_001139748.1 | ||
| NM_001146277.1 | NP_001139749.1 | |||
| NM_001146278.1 | NP_001139750.1 | |||
| NM_020792.4 | NP_065843.3 | |||
| AB037784.1 | ||||
| AK294811.1 | ||||
| AK295641.1 | ||||
| AK299422.1 | ||||
| AK304253.1 | ||||
| AK316431.1 | ||||
| BC028734.1 | AAH28734.2 | |||
| BC047588.1 | AAH47588.2 | |||
| DC306836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471060 | Chr.3:172622817 - 172682165 on Build GRCh38 | Gain |
|
| dgv8730n54 | Chr.3:172604186 - 172680078 on Build GRCh38 | Gain |
|
| dgv627n27 | Chr.3:172621962 - 172680078 on Build GRCh38 | Gain |
|
| nsv4118 | Chr.3:172638465 - 172669092 on Build GRCh38 | Insertion |
|
| esv3598669 | Chr.3:172612012 - 172681673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map