Assay Details
Target Gene Details
Entrez Gene ID: | 7325 |
Gene Name: | ubiquitin conjugating enzyme E2 E2 |
Gene Aliases: |
UBCH8 |
Location: |
Chr.3:23203007-23590805 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2E2 | NM_152653.3 | NP_689866.1 | ||
| XM_005265432.2 | XP_005265489.1 | |||
| XM_005265433.4 | XP_005265490.1 | |||
| XM_011534074.2 | 4 | 1619 | XP_011532376.1 | |
| XM_011534075.1 | XP_011532377.1 | |||
| XM_011534076.2 | XP_011532378.1 | |||
| XM_017007126.1 | XP_016862615.1 | |||
| AK057886.1 | BAB71605.1 | |||
| AK307075.1 | ||||
| BC022332.1 | AAH22332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834636 | Chr.3:23097162 - 23259952 on Build GRCh38 | Gain+Loss |
|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map