Assay Details
Target Gene Details
Entrez Gene ID: | 1238 |
Gene Name: | atypical chemokine receptor 2 |
Gene Aliases: |
CCBP2, CCR10, CCR9, CMKBR9, D6, hD6 |
Location: |
Chr.3:42809472-42867283 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACKR2 | NM_001296.4 | NP_001287.2 | ||
| AK313561.1 | ||||
| BC008816.2 | AAH08816.1 | |||
| BC011631.2 | AAH11631.1 | |||
| BC020558.1 | AAH20558.1 | |||
| BE901130.1 | ||||
| DA853159.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114068921] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map