Assay Details
Target Gene Details
Entrez Gene ID: | 8545 |
Gene Name: | CGG triplet repeat binding protein 1 |
Gene Aliases: |
CGGBP, p20-CGGBP |
Location: |
Chr.3:88051950-88149866 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CGGBP1 | NM_001195308.1 | NP_001182237.1 |
Target Gene Details
Entrez Gene ID: | 55279 |
Gene Name: | zinc finger protein 654 |
Gene Aliases: |
- |
Location: |
Chr.3:88059217-88144664 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF654 | XM_011534340.2 | XP_011532642.2 | ||
| XM_017006789.1 | XP_016862278.1 | |||
| XM_017006790.1 | XP_016862279.1 | |||
| XM_017006791.1 | XP_016862280.1 | |||
| XM_017006792.1 | XP_016862281.1 | |||
| XM_017006793.1 | XP_016862282.1 | |||
| XM_017006795.1 | XP_016862284.1 |
More Information
Set Membership: |
|

Genomic Map