Assay Details
Target Gene Details
Entrez Gene ID: | 22906 |
Gene Name: | trafficking kinesin protein 1 |
Gene Aliases: |
MILT1, OIP106 |
Location: |
Chr.3:42013802-42225890 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAK1 | XM_017005906.1 | XP_016861395.1 | ||
| XM_017005909.1 | XP_016861398.1 | |||
| XM_017005911.1 | XP_016861400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460520 | Chr.3:41918514 - 42139904 on Build GRCh38 | Gain |
|
| nsv954469 | Chr.3:42027509 - 42034308 on Build GRCh38 | Deletion |
|
| esv2760776 | Chr.3:41872696 - 42174077 on Build GRCh38 | Gain |
|
| dgv8295n54 | Chr.3:41843258 - 42142869 on Build GRCh38 | Gain |
|
| esv3595984 | Chr.3:41873454 - 42129694 on Build GRCh38 | Gain |
|
| dgv8296n54 | Chr.3:41859164 - 42073996 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map