Assay Details
Target Gene Details
Entrez Gene ID: | 55799 |
Gene Name: | calcium voltage-gated channel auxiliary subunit alpha2delta 3 |
Gene Aliases: |
HSA272268 |
Location: |
Chr.3:54122594-55074557 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Gene Details
Entrez Gene ID: | 100874237 |
Gene Name: | CACNA2D3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:54874605-54901255 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA2D3-AS1 | NR_046666.1 | |||
| AK092143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517851 | Chr.3:54375499 - 55705332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map