Assay Details
Target Gene Details
Entrez Gene ID: | 83478 |
Gene Name: | Rho GTPase activating protein 24 |
Gene Aliases: |
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP |
Location: |
Chr.4:85475131-86002670 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP24 | NM_001025616.2 | NP_001020787.2 | ||
| AK091196.1 | BAC03606.1 | |||
| BC047918.1 | AAH47918.1 | |||
| DA815489.1 | ||||
| DB109452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519588 | Chr.4:84841232 - 85670049 on Build GRCh38 | Gain |
|
| nsv594747 | Chr.4:85066879 - 85804349 on Build GRCh38 | Gain |
|
| nsv470048 | Chr.4:85077630 - 85799685 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78211312] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map