Assay Details
Target Gene Details
Entrez Gene ID: | 80306 |
Gene Name: | mediator complex subunit 28 |
Gene Aliases: |
1500003D12Rik, EG1, magicin |
Location: |
Chr.4:17614628-17625628 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED28 | NM_025205.4 | NP_079481.2 | ||
| AF317678.1 | ||||
| AF317679.1 | ||||
| AF317680.1 | AAK11563.1 | |||
| AF318059.1 | AAL26906.1 | |||
| AF358829.1 | AAK32724.1 | |||
| AK313928.1 | ||||
| BC011936.2 | AAH11936.1 | |||
| HY174042.1 |
More Information
Set Membership: |
|

Genomic Map