Assay Details
Target Gene Details
Entrez Gene ID: | 4258 |
Gene Name: | microsomal glutathione S-transferase 2 |
Gene Aliases: |
GST2, MGST-II |
Location: |
Chr.4:139665725-139754546 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST2 | NM_001204366.1 | NP_001191295.1 | ||
| NM_001204367.1 | NP_001191296.1 | |||
| NM_001204368.1 | NP_001191297.1 | |||
| NM_002413.4 | NP_002404.1 | |||
| XM_006714221.2 | XP_006714284.1 | |||
| XM_011531972.2 | XP_011530274.1 | |||
| XM_017008211.1 | XP_016863700.1 | |||
| XM_017008212.1 | XP_016863701.1 | |||
| XM_017008213.1 | XP_016863702.1 | |||
| XM_017008214.1 | XP_016863703.1 | |||
| AA122237.1 | ||||
| AK130948.1 | ||||
| AK307231.1 | ||||
| BC025416.1 | AAH25416.1 | |||
| BE312976.1 | ||||
| CR407640.1 | CAG28568.1 | |||
| DB257575.1 | ||||
| U77604.1 | AAC51768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569728 | Chr.4:139679425 - 139688643 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map