Assay Details
Target Gene Details
Entrez Gene ID: | 3426 |
Gene Name: | complement factor I |
Gene Aliases: |
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF |
Location: |
Chr.4:109731221-109802225 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFI | NM_000204.4 | NP_000195.2 | ||
| NM_001318057.1 | NP_001304986.1 | |||
| XM_005262976.1 | XP_005263033.1 | |||
| XM_006714209.1 | XP_006714272.1 | |||
| XM_006714210.3 | XP_006714273.1 | |||
| XM_011531920.1 | XP_011530222.1 | |||
| XM_017008164.1 | XP_016863653.1 | |||
| XM_017008165.1 | XP_016863654.1 | |||
| XM_017008166.1 | XP_016863655.1 | |||
| AK122686.1 | ||||
| AK290625.1 | ||||
| AK299232.1 | ||||
| BC020718.1 | AAH20718.1 | |||
| J02770.1 | AAA52455.1 | |||
| KU159429.1 | ||||
| Y00318.1 | CAA68416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4463 | Chr.4:109726575 - 109771969 on Build GRCh38 | Deletion |
|
| esv3569652 | Chr.4:109727450 - 109747513 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map