Assay Details
Target Gene Details
Entrez Gene ID: | 100885776 |
Gene Name: | UGDH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:39527839-39594707 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGDH-AS1 | NR_047679.1 | |||
| AK056558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010784 | Chr.4:39437476 - 39571758 on Build GRCh38 | Gain |
|

Genomic Map