Assay Details
Target Gene Details
Entrez Gene ID: | 4750 |
Gene Name: | NIMA related kinase 1 |
Gene Aliases: |
NY-REN-55, SRPS2, SRPS2A, SRTD6 |
Location: |
Chr.4:169393270-169612629 on Build GRCh38 |
Assay Gene Location: | Within Intron 36 |
More Information
Set Membership: |
|

Genomic Map