Assay Details
Target Gene Details
Entrez Gene ID: | 287 |
Gene Name: | ankyrin 2, neuronal |
Gene Aliases: |
ANK-2, LQT4, brank-2 |
Location: |
Chr.4:112706083-113383740 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Gene Details
Entrez Gene ID: | 102465878 |
Gene Name: | microRNA 8082 |
Gene Aliases: |
hsa-mir-8082 |
Location: |
Chr.4:113152282-113152362 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR8082 | NR_107049.1 |
More Information
Set Membership: |
|

Genomic Map