Assay Details
Target Gene Details
Entrez Gene ID: | 285540 |
Gene Name: | SEPSECS antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:25160672-25198506 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPSECS-AS1 | NR_037934.1 | |||
| AK095450.1 | ||||
| BX646650.1 |

Genomic Map