Assay Details
Target Gene Details
Entrez Gene ID: | 9508 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 3 |
Gene Aliases: |
ADAMTS-4 |
Location: |
Chr.4:72280969-72568799 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS3 | NM_014243.2 | NP_055058.2 | ||
| XM_011532421.2 | XP_011530723.1 | |||
| XM_011532422.2 | XP_011530724.1 | |||
| AB002364.1 | BAA20821.1 | |||
| BC130287.1 | ||||
| BC132735.1 |
More Information
Set Membership: |
|

Genomic Map