Assay Details
Target Gene Details
Entrez Gene ID: | 3426 |
Gene Name: | complement factor I |
Gene Aliases: |
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF |
Location: |
Chr.4:109731221-109802225 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFI | XM_011531920.1 | XP_011530222.1 | ||
| XM_017008164.1 | XP_016863653.1 | |||
| XM_017008165.1 | XP_016863654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4463 | Chr.4:109726575 - 109771969 on Build GRCh38 | Deletion |
|
| nsv595142 | Chr.4:109688325 - 109735574 on Build GRCh38 | Loss |
|
| esv3569652 | Chr.4:109727450 - 109747513 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77673871] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map