Assay Details
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM165 | NM_018475.4 | 1 | 133 | NP_060945.2 |
| NR_073070.1 | 1 | 133 | ||
| XM_011534394.2 | 1 | 123 | XP_011532696.1 | |
| AF220188.1 | AAF67653.1 | |||
| AK290663.1 | 1 | 86 | ||
| AK295289.1 | 1 | 83 | ||
| BC107582.1 | 1 | 93 | AAI07583.1 | |
| DA371259.1 | 1 | 123 | ||
| HY089602.1 | 1 | 135 |
More Information
Additional Information:
For this assay, SNP(s) [rs116687212] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map