Assay Details
Target Gene Details
Entrez Gene ID: | 90768 |
Gene Name: | uncharacterized LOC90768 |
Gene Aliases: |
- |
Location: |
Chr.4:182138658-182144515 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC90768 | NR_027107.2 | 4 | 3262 | |
| NR_125905.1 | 5 | 1309 | ||
| AK094166.1 | 1 | 778 | ||
| DA296106.1 |
Target Gene Details
Entrez Gene ID: | 55714 |
Gene Name: | teneurin transmembrane protein 3 |
Gene Aliases: |
MCOPCB9, ODZ3, TNM3, Ten-m3, ten-3 |
Location: |
Chr.4:181447613-182803024 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TENM3 | XM_017008385.1 | XP_016863874.1 | ||
| XM_017008389.1 | XP_016863878.1 | |||
| XM_017008390.1 | XP_016863879.1 | |||
| XM_017008391.1 | XP_016863880.1 | |||
| XM_017008392.1 | XP_016863881.1 | |||
| XM_017008393.1 | XP_016863882.1 | |||
| XM_017008394.1 | XP_016863883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016288 | Chr.4:179729085 - 182726257 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map