Assay Details
Target Gene Details
Entrez Gene ID: | 5860 |
Gene Name: | quinoid dihydropteridine reductase |
Gene Aliases: |
DHPR, PKU2, SDR33C1 |
Location: |
Chr.4:17486393-17512234 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QDPR | NM_000320.2 | NP_000311.2 | ||
| NM_001306140.1 | NP_001293069.1 | |||
| AK124382.1 | ||||
| AK223437.1 | BAD97157.1 | |||
| AK289773.1 | ||||
| AK296622.1 | ||||
| BC000576.2 | AAH00576.1 | |||
| BM762805.1 | ||||
| M16447.1 | AAA52305.1 | |||
| X04882.1 | CAA28571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003127 | Chr.4:17454748 - 17540540 on Build GRCh38 | Gain |
|
| nsv593781 | Chr.4:17030351 - 17591490 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73234956] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map