Assay Details
Target Gene Details
Entrez Gene ID: | 132949 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase |
Gene Aliases: |
ACSF4, LYS2, NRPS1098, NRPS998 |
Location: |
Chr.4:56337687-56387677 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161086 | Chr.4:56310907 - 56510457 on Build GRCh38 | Duplication |
|
| nsv829940 | Chr.4:56206175 - 56396564 on Build GRCh38 | Gain |
|
| dgv5213n100 | Chr.4:56076751 - 56538345 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map