Assay Details
Target Gene Details
Entrez Gene ID: | 57654 |
Gene Name: | UV stimulated scaffold protein A |
Gene Aliases: |
KIAA1530, UVSS3 |
Location: |
Chr.4:1345662-1388049 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UVSSA | NM_001317934.1 | NP_001304863.1 | ||
| NM_001317935.1 | NP_001304864.1 | |||
| NM_020894.3 | NP_065945.2 | |||
| XM_017008490.1 | XP_016863979.1 | |||
| XM_017008491.1 | XP_016863980.1 | |||
| XM_017008492.1 | XP_016863981.1 | |||
| XM_017008493.1 | XP_016863982.1 | |||
| XM_017008494.1 | XP_016863983.1 | |||
| XM_017008495.1 | XP_016863984.1 | |||
| XM_017008498.1 | XP_016863987.1 | |||
| XM_017008500.1 | XP_016863989.1 | |||
| AB040963.1 | ||||
| AK292661.1 | ||||
| BC110331.1 | AAI10332.1 | |||
| BC140901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950332 | Chr.4:1364213 - 1367912 on Build GRCh38 | Deletion |
|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss |
|
| nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77128276,rs80140145] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map