Assay Details
Target Gene Details
Entrez Gene ID: | 255743 |
Gene Name: | nephronectin |
Gene Aliases: |
EGFL6L, POEM |
Location: |
Chr.4:105895440-105971671 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPNT | NM_001033047.2 | NP_001028219.1 | ||
| NM_001184690.1 | NP_001171619.1 | |||
| NM_001184691.1 | NP_001171620.1 | |||
| NM_001184692.1 | NP_001171621.1 | |||
| NM_001184693.1 | NP_001171622.1 | |||
| XM_005262888.4 | XP_005262945.1 | |||
| XM_005262890.1 | XP_005262947.1 | |||
| XM_011531820.2 | XP_011530122.1 | |||
| XM_011531822.2 | XP_011530124.1 | |||
| XM_011531823.2 | XP_011530125.1 | |||
| XM_011531824.2 | XP_011530126.1 | |||
| XM_011531825.2 | XP_011530127.1 | |||
| XM_017007984.1 | XP_016863473.1 | |||
| AK290029.1 | ||||
| AK295772.1 | ||||
| AK302477.1 | ||||
| AK304279.1 | ||||
| AK304721.1 | ||||
| AL832465.1 | ||||
| AY358336.1 | AAQ88702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
| nsv1014993 | Chr.4:105874862 - 106059570 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map