Assay Details
Target Gene Details
Entrez Gene ID: | 81608 |
Gene Name: | factor interacting with PAPOLA and CPSF1 |
Gene Aliases: |
FIP1, Rhe, hFip1 |
Location: |
Chr.4:53377572-53462611 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569422 | Chr.4:53379608 - 53385331 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map