Assay Details
Target Gene Details
Entrez Gene ID: | 10886 |
Gene Name: | neuropeptide FF receptor 2 |
Gene Aliases: |
GPR74, HLWAR77, NPFF2, NPGPR |
Location: |
Chr.4:72031804-72148201 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPFFR2 | NM_001144756.1 | NP_001138228.1 | ||
| NM_004885.2 | NP_004876.2 | |||
| NM_053036.2 | NP_444264.1 | |||
| XM_011531554.2 | XP_011529856.1 | |||
| AF119815.1 | AAD22047.1 | |||
| AF236083.1 | AAK58513.1 | |||
| AF257210.1 | AAF87078.1 | |||
| AJ311393.1 | CAC85427.1 | |||
| AK290937.1 | ||||
| AK308069.1 | ||||
| BC101636.1 | ||||
| BC113713.1 |
More Information
Set Membership: |
|

Genomic Map