Assay Details
Target Gene Details
Entrez Gene ID: | 8821 |
Gene Name: | inositol polyphosphate-4-phosphatase type II B |
Gene Aliases: |
- |
Location: |
Chr.4:142023160-142846535 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPP4B | XM_017008794.1 | 27 | 4844 | XP_016864283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830096 | Chr.4:141945946 - 142102608 on Build GRCh38 | Gain |
|
| nsv1027781 | Chr.4:142022332 - 142385414 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map