Assay Details
Target Gene Details
Entrez Gene ID: | 10563 |
Gene Name: | C-X-C motif chemokine ligand 13 |
Gene Aliases: |
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13 |
Location: |
Chr.4:77511753-77611834 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL13 | NM_006419.2 | NP_006410.1 | ||
| AF044197.1 | AAC14402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829975 | Chr.4:77505056 - 77644044 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72861963] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map