Assay Details
Target Gene Details
Entrez Gene ID: | 255130 |
Gene Name: | IGFBP7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:57109762-57205299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFBP7-AS1 | NR_034081.1 | |||
| BG425784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600596 | Chr.4:57137231 - 57156271 on Build GRCh38 | Loss |
|
| nsv594207 | Chr.4:57121267 - 57232388 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67746677] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map