Assay Details
Target Gene Details
Entrez Gene ID: | 100507053 |
Gene Name: | uncharacterized LOC100507053 |
Gene Aliases: |
- |
Location: |
Chr.4:99088857-99301356 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100507053 | NR_037884.1 | |||
BX647987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1947060 | Chr.4:99101685 - 99102418 on Build GRCh38 | Deletion |
![]() |
esv5687 | Chr.4:99101614 - 99102489 on Build GRCh38 | Loss |
![]() |
nsv1002927 | Chr.4:99031321 - 99227533 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2569136 | Chr.4:99101108 - 99102760 on Build GRCh38 | Deletion |
![]() |
esv3575764 | Chr.4:99092644 - 99102819 on Build GRCh38 | Gain |
![]() |
