Assay Details
Target Gene Details
Entrez Gene ID: | 133121 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 6 |
Gene Aliases: |
NPP6 |
Location: |
Chr.4:184088706-184217961 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP6 | NM_153343.3 | NP_699174.1 | ||
| AK057370.1 | BAB71455.1 | |||
| AY358676.1 | AAQ89039.1 | |||
| BC035035.2 | AAH35035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521658 | Chr.4:184057338 - 184121636 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77116912,rs80188473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map