Assay Details
Target Gene Details
Entrez Gene ID: | 23328 |
Gene Name: | SAM and SH3 domain containing 1 |
Gene Aliases: |
SH3D6A, dJ323M4, dJ323M4.1 |
Location: |
Chr.6:148193440-148552049 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SASH1 | XM_017010599.1 | XP_016866088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520615 | Chr.6:147807005 - 148369628 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map