Assay Details
Target Gene Details
Entrez Gene ID: | 80177 |
Gene Name: | myc target 1 |
Gene Aliases: |
MTLC |
Location: |
Chr.6:152697895-152747171 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCT1 | NM_025107.2 | NP_079383.2 | ||
| XM_011536149.2 | XP_011534451.1 | |||
| XM_011536150.2 | XP_011534452.1 | |||
| XM_011536151.2 | XP_011534453.1 | |||
| XM_011536152.2 | XP_011534454.1 | |||
| XM_017011319.1 | XP_016866808.1 | |||
| AF527367.1 | AAM88866.1 | |||
| AK021612.1 | ||||
| AK024922.1 | BAB15035.1 | |||
| AL834500.1 | CAD39158.3 | |||
| BC022556.1 | AAH22556.2 | |||
| BC128463.1 | ||||
| BC128464.1 | ||||
| DA863771.1 | ||||
| GU997693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5546 | Chr.6:152700158 - 152718204 on Build GRCh38 | Deletion |
|
| nsv1117675 | Chr.6:152712655 - 152735303 on Build GRCh38 | Deletion |
|

Genomic Map