Assay Details
Target Gene Details
Entrez Gene ID: | 3096 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 1 |
Gene Aliases: |
CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A |
Location: |
Chr.6:12002096-12212034 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIVEP1 | NM_002114.3 | NP_002105.3 | ||
| XM_011514546.2 | XP_011512848.1 | |||
| XM_011514547.2 | XP_011512849.1 | |||
| XM_011514548.2 | XP_011512850.1 | |||
| XM_011514549.2 | XP_011512851.1 | |||
| XM_011514550.2 | XP_011512852.1 | |||
| XM_011514551.2 | XP_011512853.1 | |||
| XM_011514552.2 | XP_011512854.1 | |||
| XM_011514553.1 | XP_011512855.1 | |||
| XM_017010800.1 | XP_016866289.1 | |||
| XM_017010801.1 | XP_016866290.1 | |||
| XM_017010802.1 | XP_016866291.1 | |||
| XM_017010803.1 | XP_016866292.1 | |||
| XM_017010804.1 | XP_016866293.1 | |||
| AI033710.1 | ||||
| BC140816.1 | ||||
| X51435.1 | CAA35798.1 |
More Information
Set Membership: |
|

Genomic Map