Assay Details
Target Gene Details
Entrez Gene ID: | 8876 |
Gene Name: | vanin 1 |
Gene Aliases: |
HDLCQ8, Tiff66 |
Location: |
Chr.6:132680858-132714055 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VNN1 | NM_004666.2 | NP_004657.2 | ||
| AJ132099.1 | CAA10568.1 | |||
| AK290425.1 | ||||
| AK314265.1 | ||||
| BC096265.3 | AAH96265.1 | |||
| BC096266.3 | AAH96266.1 | |||
| BC096267.3 | AAH96267.1 | |||
| BC096268.1 | AAH96268.1 | |||
| U39664.1 | AAF21453.1 |
More Information
Set Membership: |
|

Genomic Map