Assay Details
Target Gene Details
Entrez Gene ID: | 387357 |
Gene Name: | thymocyte selection associated |
Gene Aliases: |
C6orf190, C6orf207, GASP, SPOT, TSEPA |
Location: |
Chr.6:127697191-127918631 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THEMIS | NM_001010923.2 | NP_001010923.1 | ||
| NM_001164685.1 | NP_001158157.1 | |||
| NM_001164687.1 | NP_001158159.1 | |||
| NM_001318531.1 | NP_001305460.1 | |||
| XM_011535813.2 | XP_011534115.1 | |||
| XM_011535814.2 | XP_011534116.1 | |||
| XM_011535816.1 | XP_011534118.1 | |||
| XM_017010849.1 | XP_016866338.1 | |||
| AK094863.1 | ||||
| AK097903.1 | BAC05194.1 | |||
| AK124031.1 | ||||
| AK128377.1 | ||||
| AK292046.1 | ||||
| BC130516.1 | ||||
| BX640890.1 | CAE45941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830799 | Chr.6:127711955 - 127873366 on Build GRCh38 | Loss |
|
| esv3610728 | Chr.6:127723211 - 127729005 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map