Assay Details
Target Gene Details
Entrez Gene ID: | 387357 |
Gene Name: | thymocyte selection associated |
Gene Aliases: |
C6orf190, C6orf207, GASP, SPOT, TSEPA |
Location: |
Chr.6:127697191-127918631 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THEMIS | XM_011535813.2 | XP_011534115.1 |
More Information
Set Membership: |
|

Genomic Map