Assay Details
Target Gene Details
Entrez Gene ID: | 8504 |
Gene Name: | peroxisomal biogenesis factor 3 |
Gene Aliases: |
PBD10A, TRG18 |
Location: |
Chr.6:143450781-143490616 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEX3 | NM_003630.2 | NP_003621.1 | ||
| AB035307.1 | BAA97993.1 | |||
| AJ001625.1 | CAA04879.1 | |||
| AJ131389.1 | CAA10362.1 | |||
| AK300907.1 | ||||
| AY277600.1 | AAQ18039.1 | |||
| BC014551.1 | AAH14551.1 | |||
| BC015506.1 | AAH15506.1 | |||
| BP377713.1 | ||||
| CR542062.1 | CAG46859.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78684160] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map