Assay Details
Target Gene Details
Entrez Gene ID: | 7754 |
Gene Name: | zinc finger protein 204, pseudogene |
Gene Aliases: |
ZNF184-Lp, ZNF204, ZNF315P, b24o18.1 |
Location: |
Chr.6:27357823-27375374 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF204P | NR_024553.1 | |||
| DA147897.1 | ||||
| DB031764.1 |

Genomic Map