Assay Details
Target Gene Details
Entrez Gene ID: | 80177 |
Gene Name: | myc target 1 |
Gene Aliases: |
MTLC |
Location: |
Chr.6:152697895-152747171 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCT1 | XM_017011319.1 | XP_016866808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1117675 | Chr.6:152712655 - 152735303 on Build GRCh38 | Deletion |
|

Genomic Map