Assay Details
Target Gene Details
Entrez Gene ID: | 7754 |
Gene Name: | zinc finger protein 204, pseudogene |
Gene Aliases: |
ZNF184-Lp, ZNF204, ZNF315P, b24o18.1 |
Location: |
Chr.6:27357823-27375374 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF204P | NR_024553.1 | |||
| DA147897.1 | ||||
| DB031764.1 |
Target Gene Details
Entrez Gene ID: | 346157 |
Gene Name: | zinc finger protein 391 |
Gene Aliases: |
dJ153G14.3 |
Location: |
Chr.6:27374635-27403908 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF391 | NM_001322288.1 | 1 | 389 | NP_001309217.1 |
| XM_006715086.2 | XP_006715149.1 | |||
| XM_011514569.1 | XP_011512871.1 | |||
| BG722372.1 | 1 | 394 | ||
| DB060105.1 | 1 | 409 |
More Information
Set Membership: |
|

Genomic Map