Assay Details
Target Gene Details
Entrez Gene ID: | 114792 |
Gene Name: | kelch like family member 32 |
Gene Aliases: |
BKLHD5, KIAA1900, UG0030H05, dJ21F7.1 |
Location: |
Chr.6:96898083-97142949 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL32 | NM_001323258.1 | 1 | 1022 | NP_001310187.1 |
| HY013072.1 |
More Information
Set Membership: |
|

Genomic Map